Learn More
Invitrogen™ beta-2 Defensin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107328
Description
Product is supplied at 1 mL (100 μL/mL). Application Notes: Western Blot - Test was performend under reduced sample treatment using SDS-Page. The band size is 25kDa for recombinant protein with tag and 5 kDa for the native protein. IHC-P- Tissue sections were pretreated with high temperature citrate and fixed in formalin. PBS was used as a negative control.
Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The alpha-defensins are distinguished from the beta-defensins by the pairing of their three disulfide bonds. To date, six human beta-defensins have been identified; BD-1, BD-2, BD-3, BD-4, BD-5 and BD-6. beta-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they can act as chemoattractants towards immature dendritic cells and memory T cells. The beta-defensin proteins are expressed as the C-terminal portion of precursors, and are released by proteolytic cleavage of a signal sequence and, in some cases, a propeptide sequence. Beta-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds.
Specifications
| beta-2 Defensin | |
| Polyclonal | |
| Unconjugated | |
| DEFB4A | |
| BD-2; beta defensin 2; beta defensin-2; Beta-defensin 2; Beta-defensin 4A; DEFB102; DEFB2; DEFB-2; DEFB4; DEFB4A; DEFB4B; Defensin beta 2; defensin beta 4A; Defensin beta2; defensin, beta 2; defensin, beta 4; defensin, beta 4A; Defensin-beta2; HBD-2; SAP1; Skin-antimicrobial peptide 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1673 | |
| 4°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 100 μg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| O15263 | |
| DEFB4A | |
| CHPVFCPRRYKQIGTCGL. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.