missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-2 Adrenergic R/ADRB2 Mouse anti-Human, Clone: 13D6, Novus Biologicals™
Shop All Bio Techne ProductsDescription
beta-2 Adrenergic R/ADRB2 Monoclonal antibody specifically detects beta-2 Adrenergic R/ADRB2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunoprecipitation
Specifications
Specifications
| Antigen | beta-2 Adrenergic R/ADRB2 |
| Applications | Western Blot, ELISA, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 13D6 |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Formulation | PBS |
| Gene Alias | ADRBR, adrenergic, beta-2-, receptor, surface, B2ARADRB2R, BAR, beta-2 adrenergic receptor, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?