missing translation for 'onlineSavingsMsg'
Learn More

beta-2 Adrenergic R/ADRB2 Antibody (R11/E1), DyLight 594, Novus Biologicals™

Product Code. 18385363 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18385363 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18385363 Supplier Novus Biologicals Supplier No. NBP312006DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

beta-2 Adrenergic R/ADRB2 Monoclonal antibody specifically detects beta-2 Adrenergic R/ADRB2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-2 Adrenergic R/ADRB2
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone R11/E1
Conjugate DyLight 594
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ADRBR, adrenergic, beta-2-, receptor, surface, B2ARADRB2R, BAR, beta-2 adrenergic receptor, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor
Host Species Rabbit
Immunogen This antibody was raised by immunizing mice with beta-2 adrenergic receptor.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, GPCR, Immunology, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 154
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.