missing translation for 'onlineSavingsMsg'
Learn More

beta-2 Adrenergic R/ADRB2 Antibody (6H8), Novus Biologicals™

Product Code. 18374303 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374303 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18374303 Supplier Novus Biologicals Supplier No. NBP311204

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta-2 Adrenergic R/ADRB2 Monoclonal specifically detects beta-2 Adrenergic R/ADRB2 in Human, Rat, Guinea Pig samples. It is validated for Flow Cytometry, Functional, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-2 Adrenergic R/ADRB2
Applications Flow Cytometry, Functional Assay, Immunoassay
Classification Monoclonal
Clone 6H8
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry, Functional, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias ADRBR, adrenergic, beta-2-, receptor, surface, B2ARADRB2R, BAR, beta-2 adrenergic receptor, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor
Host Species Mouse
Immunogen Free peptide Beta2-H19C
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Asthma, GPCR, Immunology, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 154
Target Species Human, Rat, Guinea Pig
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.