Learn More
beta-2 Adrenergic R/ADRB2 Antibody (4C11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26521-100ul
 View more versions of this product
Product Code. 30043055
  • £406.00 / 100µL
Description
beta-2 Adrenergic R/ADRB2 Monoclonal antibody specifically detects beta-2 Adrenergic R/ADRB2 in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
beta-2 Adrenergic R/ADRB2
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry
4C11
ELISA, Immunohistochemistry 1:50-1:200
ADRBR, adrenergic, beta-2-, receptor, surface, B2ARADRB2R, BAR, beta-2 adrenergic receptor, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor
A synthesized peptide derived from Human beta-2 Adrenergic R/ADRB2 [UniProt P07550]
100 μL
Asthma, GPCR, Immunology, Neuroscience, Phospho Specific, Signal Transduction
154
Store at -20 to -70C. Avoid freeze-thaw cycles.