missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta 2 Adaptin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56481
This item is not returnable.
View return policy
Description
Beta 2 Adaptin Polyclonal specifically detects Beta 2 Adaptin in Human samples. It is validated for Western Blot.
Specifications
| Beta 2 Adaptin | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Adapter-related protein complex 2 beta subunit, Adaptor protein complex AP-2 subunit beta, adaptor-related protein complex 2, beta 1 subunit, ADTB2adaptin, beta 2 (beta), AP105B, AP2-BETA, Beta-2-adaptin, Beta-adaptin, CLAPB1AP-2 complex subunit beta, Clathrin assembly protein complex 2 beta large chain, clathrin-associated/assembly/adaptor protein, large, beta 1, DKFZp781K0743, Plasma membrane adaptor HA2/AP2 adaptin beta subunit | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 163 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P63010 | |
| AP2B1 | |
| Synthetic peptides corresponding to AP2B1(adaptor-related protein complex 2, beta 1 subunit) The peptide sequence was selected from the middle region of AP2B1. Peptide sequence SETQELVQQVLSLATQDSDNPDLRDRGYIYWRLLSTDPVTAKEVVLSEKP. | |
| 100 μL | |
| Primary | |
| This product is specific to Subunit or Isoform: beta. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction