missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-1,3-N-Acetylglucosaminyltransferase 4/B3GNT4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92274-0.1ml
This item is not returnable.
View return policy
Description
Beta-1,3-N-Acetylglucosaminyltransferase 4/B3GNT4 Polyclonal antibody specifically detects Beta-1,3-N-Acetylglucosaminyltransferase 4/B3GNT4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Beta-1,3-N-Acetylglucosaminyltransferase 4/B3GNT4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| B3GN-T4, beta-1,3-Gn-T4, Beta-1,3-N-acetylglucosaminyltransferase 4, beta-1,3-N-acetylglucosaminyltransferase bGn-T4, beta3Gn-T4, BGnT-4, EC 2.4.1.-, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 165-305 of human B3GNT4 (NP_110392.1). APPAQLLAYESREFDDILQWDFTEDFFNLTLKELHLQRWVVAACPQAHFMLKGDDDVFVHVPNVLEFLDGWDPAQDLLVGDVIRQALPNRNTKVKYFIPPSMYRATHYPPYAGGGGYVMSRATVRRLQAIMEDAELFPIDD | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79369 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction