missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97144-50ul
This item is not returnable.
View return policy
Description
Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 Polyclonal antibody specifically detects Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| b3Gal-T7, B3GALT7, B3GNT, B3GNT1, B3GN-T1, B3GN-T2, B3GNT-2, Beta-1,3-galactosyltransferase 7, Beta-1,3-GalTase 7, beta-1,3-Gn-T1, Beta-1,3-Gn-T2, Beta-1,3-N-acetylglucosaminyltransferase 1, Beta-1,3-N-acetylglucosaminyltransferase 2, beta-1,3-N-acetylglucosaminyltransferase bGnT-1, beta-1,3-N-acetylglucosaminyltransferase bGnT-2, Beta3GalT7, Beta3Gal-T7, BETA3GNT, Beta3Gn-T1, beta3Gn-T2, Beta-3-Gx-T7, BGnT-1, BGnT-2, BGNT2, EC 2.4.1.-, UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 7, UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 7, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2 | |
| Produced in rabbits immunized with E. coli-derived Human Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 fragment. | |
| 50 μg | |
| Cell Cycle and Replication | |
| 10678 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur