missing translation for 'onlineSavingsMsg'
Learn More

beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody (CL9500), Novus Biologicals™

Mouse Monoclonal Antibody

£317.00 - £473.00

Specifications

Antigen beta-1,3-Glucuronyltransferase 1/B3GAT1
Clone CL9500
Dilution Western Blot 1 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18606654
View Documents
Novus Biologicals
NBP3-21171-25ul
25 μg
£317.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18682805
View Documents
Novus Biologicals
NBP3-21171-100ul
100 μg
£473.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

beta-1,3-Glucuronyltransferase 1/B3GAT1 Monoclonal antibody specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

beta-1,3-Glucuronyltransferase 1/B3GAT1
Western Blot 1 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Monoclonal
Purified
RUO
Human
Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase
This antibody was developed against a Synthetic Peptide of Q9P2W7. Actual immunogen sequence is proprietary.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
CL9500
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer, Immunology
PBS, pH 7.2, 40% glycerol
27087
IgG1
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.