missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody (CL9499) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21170JF549
This item is not returnable.
View return policy
Description
beta-1,3-Glucuronyltransferase 1/B3GAT1 Monoclonal antibody specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| beta-1,3-Glucuronyltransferase 1/B3GAT1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase | |
| This antibody was developed using a synthetic peptide derived from Q9P2W7, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Immunology | |
| 27087 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CL9499 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction