Learn More
beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody (CL9499) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21170F
Product Code. 30307747
  • £558.00 / 0.10mL
Description
beta-1,3-Glucuronyltransferase 1/B3GAT1 Monoclonal antibody specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
beta-1,3-Glucuronyltransferase 1/B3GAT1
Monoclonal
FITC
Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase
This antibody was developed using a synthetic peptide derived from Q9P2W7, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Immunology
27087
Store at 4⪚C in the dark.
IgG2a
Western Blot, Immunocytochemistry/Immunofluorescence
CL9499
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified