missing translation for 'onlineSavingsMsg'
Learn More

beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody (3B8B10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£161.00 - £437.00

Specifications

Antigen beta-1,3-Glucuronyltransferase 1/B3GAT1
Clone 3B8B10
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18258791
View Documents
Novus Biologicals
NBP2-61708
0.1 mL
£437.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18654098
View Documents
Novus Biologicals
NBP2-61708-0.025ml
0.025 mL
£161.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

beta-1,3-Glucuronyltransferase 1/B3GAT1 Monoclonal specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

beta-1,3-Glucuronyltransferase 1/B3GAT1
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Unconjugated
Mouse
Human, Mouse
27087
Purified recombinant fragment of human B3GAT1 (AA: 193-334) expressed in E. Coli.
Primary
38.2 kDa
3B8B10
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
Monoclonal
Purified
Cancer, Immunology
Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase
B3GAT1
IgG1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.