missing translation for 'onlineSavingsMsg'
Learn More

BET1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

£359.00

Specifications

Antigen BET1
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18332875
View Documents
Novus Biologicals
NBP3-10803-100UL
100 μg
£359.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BET1 Polyclonal specifically detects BET1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

BET1
Western Blot
Unconjugated
Rabbit
Human
Bet1 (S. cerevisiae) homolog, BET1 homolog, BET1 homolog (S. cerevisiae), Bet1p homolog, blocked early in transport 1 homolog (S. cerevisiae), DKFZp781C0425, Golgi vesicular membrane trafficking protein p18, Golgi vesicular membrane-trafficking protein p18, HBET1
The immunogen is a synthetic peptide directed towards the N terminal region of human BET1 (NP_005859.1). Peptide sequence PPGNYGNYGYANSGYSACEEENERLTESLRSKVTAIKSLSIEIGHEVKTQ
Affinity purified
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
10282
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.