missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BET1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112368
This item is not returnable.
View return policy
Description
BET1 Polyclonal Antibody for Western Blot, ELISA
Specifications
| BET1 | |
| Polyclonal | |
| Unconjugated | |
| BET1 | |
| BET1; Bet1 golgi vesicular membrane trafficking protein; BET1 homolog; Bet1p homolog; blocked early in transport 1 homolog; Golgi vesicular membrane trafficking protein p18; golgi vesicular membrane-trafficking protein p18; hBET1; rBET1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 10282, 29631 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O15155, Q62896 | |
| BET1 | |
| Recombinant Human BET1 homolog protein (1-83AA). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction