missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BET-BAY 002 (S enantiomer), MedChemExpress
Description
BET-BAY 002 S enantiomer is the S-enantiomer of BET-BAY 002. BET-BAY 002 is a BET inhibitor.
Specifications
Specifications
| Chemical Name or Material | BET-BAY 002 (S enantiomer) |
| Physical Form | Solid |
| CAS | 2070009-49-1 |
| Color | Yellow |
| Quantity | 2 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H18ClN5O |
| For Use With (Application) | Neuroscience-Neuromodulation |
| SMILES | CC1=NN=C2N1C3=CC=CC=C3C(C4=CC=C(Cl)C=C4)=C[C@@H]2CC5=NN=C(C)O5 |
| Molecular Weight (g/mol) | 403.86 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?