missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bestrophin 4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568693
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 79%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
This gene is a member of the bestrophin gene family of anion channels. Bestrophin genes share a similar gene structure with highly conserved exon-intron boundaries, but with distinct 3' ends. Bestrophins are transmembrane proteins that contain a homologous region rich in aromatic residues, including an invariant arg-phe-pro motif. Mutation in one of the family members (bestrophin 1) is associated with vitelliform macular dystrophy. The bestrophin 4 gene is predominantly expressed in the colon.
Specifications
| Bestrophin 4 | |
| Polyclonal | |
| Unconjugated | |
| BEST4 | |
| BEST4; bestrophin 4; bestrophin-4; si:ch211-106k21.4; similar to vitelliform macular dystrophy 2-like 2; vitelliform macular dystrophy 2-like 2; vitelliform macular dystrophy 2-like protein 2; VMD2L2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 266675 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1.0 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NFU0 | |
| BEST4 | |
| synthetic peptide directed towards the N terminal region of human Bestrophin 4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction