missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bestrophin 3 Antibody (OTI2H3), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72407FR
This item is not returnable.
View return policy
Description
Bestrophin 3 Monoclonal specifically detects Bestrophin 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Bestrophin 3 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| BEST3 | |
| Full length recombinant protein of human Bestrophin 3 (NP_689652) produced in HEK293T cells. | |
| 0.1 mL | |
| 144453.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI2H3 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| bestrophin 3, bestrophin-3, MGC13168, MGC40411, vitelliform macular dystrophy 2-like 3, VMD2L3Vitelliform macular dystrophy 2-like protein 3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction