missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bestrophin-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21910-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the bestrophin gene family. This small gene family is characterized by proteins with a highly conserved N-terminus with four to six transmembrane domains. Bestrophins may form chloride ion channels or may regulate voltage-gated L-type calcium-ion channels. Bestrophins are generally believed to form calcium-activated chloride-ion channels in epithelial cells but they have also been shown to be highly permeable to bicarbonate ion transport in retinal tissue. Mutations in this gene are responsible for juvenile-onset vitelliform macular dystrophy (VMD2), also known as Best macular dystrophy, in addition to adult-onset vitelliform macular dystrophy (AVMD) and other retinopathies. Alternative splicing results in multiple variants encoding distinct isoforms.Specifications
| Bestrophin-1 | |
| Polyclonal | |
| Unconjugated | |
| BEST1 | |
| ARB, BEST, BEST1, bestrophin 1, Bestrophin-1, BMD, TU15B, VMD2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7439 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.51 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O76090 | |
| BEST1 | |
| Bestrophin-1 Fusion Protein Ag15129 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction