Learn More
Invitrogen™ Bestrophin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120943
Description
Positive test controls include: THP-1, BT-474, HeLa, Mouse heart, Mouse lung. The target is usually found in the following locations: Basolateral cell membrane, Cell membrane, Multi-pass membrane protein. Immunogen sequence: QPNQEDEEDA HAGIIGRFLG LQSHDHHPPR ANSRTKLLWP KRESLLHEGL PKNHKAAKQN VRGQEDNKAW KLKAVDAFKS APLYQRPGYY SAPQTPLSPT PMFFPLEPSA PSKLHSVTGI DTKDKSLKTV SSGAKKSFEL LSESDGALME HPEVSQVRRK TVEFNLTDMP EIPENHLKEP LEQSPTNIHT TLKDHMDPYW ALENRDEAHS.
The retinal pigment epithelium (RPE) and choroid represent a differentiated system of the eye that sustains normal retinal health and function. Best vitelliform macular dystrophy, known as Best disease, is an early-onset autosomal dominant condition in which accumulation of lipofuscin-like material within and beneath the RPE leads to progressive loss of central vision. The lipofuscin-like material in the macular area appears as a yellow mass like the yolk of an egg that later becomes darker and irregular in color, a process known as 'scrambling the egg'. Best disease is frequently a reflection of mutations in the Bestrophin gene, which encodes a protein containing four putative transmembrane domains and localizes to the basolateral plasma membrane of RPE cells. Human Bestrophin forms oligomeric chloride channels that are sensitive to intracellular calcium. Missense mutations at the Bestrophin locus reduces or abolishes Bestrophin protein mediated membrane current. The human Bestrophin gene encodes a 585 amino acid protein.
Specifications
| Bestrophin 1 | |
| Polyclonal | |
| Unconjugated | |
| BEST1 | |
| ARB; BEST; BEST 1; Best disease; best macular dystrophy; BEST1; Best1V1Delta2; bestrophin; bestrophin 1; Bestrophin1; Bestrophin-1; BMD; Bmd1; LOW QUALITY PROTEIN: bestrophin-1; mBest1; RP50; TU15B; vitelliform macular dystrophy (Best disease, bestrophin); vitelliform macular dystrophy 2 homolog; vitelliform macular dystrophy protein 2; Vitelliform macular dystrophy protein 2 homolog; VMD 2; Vmd2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 24115, 7439 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O76090, O88870 | |
| BEST1 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.