missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bestatin, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Bestatin |
| CAS | 58970-76-6 |
| Molecular Formula | C16H24N2O4 |
| Quantity | 100 mg |
| Synonym | [S-(R*,S*)]-N-(3-amino-2-hydroxy-1-oxo-4-phenylbutyl)-L-leucine, (-)-Bestatin, NK 421, NSC 265489, Ubenimex |
| InChI Formula | InChI=1S/C16H24N2O4/c1-10(2)8-13(16(21)22)18-15(20)14(19)12(17)9-11-6-4-3-5-7-11/h3-7,10,12-14,19H,8-9,17H2,1-2H3,(H,18,20)(H,21,22)/t12-,13+,14+/m1/s1 |
| SMILES | O=C(O)[C@H](CC(C)C)NC([C@@H](O)[C@H](N)CC1=CC=CC=C1)=O |
| IUPAC Name | (2S)-2-[[(2S,3R)-3-amino-2-hydroxy-4-phenylbutanoyl]amino]-4-methylpentanoic acid |
| Molecular Weight (g/mol) | 308.37 |
| Formula Weight | 308.1736 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction