missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Bestatin

Product Code. 18024842
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18024842 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18024842 Supplier Tocris Bioscience™ Supplier No. 1956/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Aminopeptidase inhibitor

Aminopeptidase inhibitor (Ki = 0.001 - 90 mM); inhibits enkephalin metabolism and leukotriene A4 hydrolase. Inhibits tumor cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

CAS 58970-76-6
Quantity 10 mg
Molecular Formula C16H24N2O4
Formula Weight 308.38
Purity >99%
Molecular Weight (g/mol) 308.38
Chemical Name or Material Bestatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.