missing translation for 'onlineSavingsMsg'
Learn More

Bestatin, 100%, MP Biomedicals™

Product Code. 11493991 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493991 1 mg 1mg
11414001 10 mg 10mg
11424001 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493991 Supplier MP Biomedicals Supplier No. 0215284401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Identificatori chimici

CAS 58970-76-6
Molecular Formula C16H24N2O4
Molecular Weight (g/mol) 308.38
MDL Number MFCD00083262
InChI Key VGGGPCQERPFHOB-MCIONIFRSA-N
Synonym bestatin, ubenimex, ubenimexum latin, ubenimex inn:jan, bestatin;ubenimex, s-2-2s,3r-3-amino-2-hydroxy-4-phenylbutanamido-4-methylpentanoic acid, ubenimex bestatin, chembl29292, 2s,3r-3-amino-2-hydroxy-4-phenylbutanoyl-l-leucine, 3-r-amino-2-s-hydroxy-4-phenylbutanoyl-s-leucine
PubChem CID 72172
IUPAC Name (2R)-2-[(2S,3R)-3-amino-2-hydroxy-4-phenylbutanamido]-4-methylpentanoic acid
SMILES CC(C)C[C@@H](NC(=O)[C@@H](O)[C@H](N)CC1=CC=CC=C1)C(O)=O

Specifica

Color White
Assay Percent Range 1
Quantity 1 mg
Formula Weight 308.4
Physical Form Powder
Chemical Name or Material Bestatin
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.