missing translation for 'onlineSavingsMsg'
Learn More

BES (White Crystalline Powder), Fisher BioReagents

Product Code. p-24674
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11448063

Brand: Fisher BioReagents BP501100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Used to make BES buffer with a useful pH range of 6.4-7.8 for biological applications
  • Literature describes BES buffer as preferred buffer for higher transfection efficiency of plasmid DNA
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10191-18-1
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00007533
InChI Key AJTVSSFTXWNIRG-UHFFFAOYSA-N
Synonym N, N-bis (2-Hydroxyethyl)-2-Amino-Ethanesulfonic Acid, N, N-bis (2-Hydroxyethyl) Taurine
PubChem CID 73243
ChEBI CHEBI:39045
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
SMILES OCCN(CCO)CCS(O)(=O)=O

Specifications

Melting Point 152°C
Color White
pH 2.5 to 5.0
Physical Form Solid
Quantity 100 g
DNase DNase free
Assay Percent Range ≥99 %
Loss on Drying 1% max.
Packaging Poly Bottle
Protease Protease free
Formula Weight 213.25
Percent Purity ≥99%
Chemical Name or Material BES
Show More Show Less
compliance-icons
Product Identifier
  • BES
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.