missing translation for 'onlineSavingsMsg'
Learn More

BES, 99+%, MP Biomedicals™

Product Code. 11374149 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374149 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11374149 Supplier MP Biomedicals Supplier No. 0210092705
 Restricted Item
Estimated Shipment: 08-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 10191-18-1
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99+%
Molecular Formula C6H15NO5S
MDL Number MFCD00007533
Synonym n,n-bis 2-hydroxyethyl-2-aminoethanesulfonic acid, bes, ethanesulfonic acid, 2-bis 2-hydroxyethyl amino, 2-bis 2-hydroxyethyl amino ethanesulfonic acid, n,n-bis 2-hydroxyethyl taurine, n,n-bis hydroxyethyl-2-aminoethanesulfonic acid, bes buffering agent, taurine, n,n-bis 2-hydroxyethyl, 2-bis 2-hydroxyethyl amino-ethanesulfonicacid
InChI Key AJTVSSFTXWNIRG-UHFFFAOYSA-N
SMILES OCCN(CCO)CCS(O)(=O)=O
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
Molecular Weight (g/mol) 213.25
PubChem CID 73243
ChEBI CHEBI:39045
Formula Weight 213.3
Chemical Name or Material BES
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.