missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BES, 99+%, for biochemistry

Product Code. 10616962 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10616962 100 g Glass bottle 100g
10082210 25 g Glass bottle 25g
11985271 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10616962 Supplier Thermo Scientific Chemicals Supplier No. 172581000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10191-18-1 | C6H15NO5S | 213.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10191-18-1
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00007533
InChI Key AJTVSSFTXWNIRG-UHFFFAOYSA-N
Synonym N, N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid
PubChem CID 73243
ChEBI CHEBI:39045
SMILES OCCN(CCO)CCS(O)(=O)=O

Specifications

Absorbance 0.095 max. (1 M in H2O) at 260nm
Melting Point 152.0°C to 154.0°C
CAS Min % 99.0
CAS Max % 100.0
Color Beige to Brown
Physical Form Powder
Quantity 100 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Loss on Drying 0.5 % max. (100°C, 2 hrs)
Linear Formula (HOCH2CH2)2NCH2CH2SO3H
Packaging Glass bottle
Solubility Information Solubility in water: soluble.
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
Formula Weight 213.25
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material BES
Show More Show Less
Product Identifier
  • BES

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.