missing translation for 'onlineSavingsMsg'
Learn More

BES, 99+%, for biochemistry, ACROS Organics™

Brand:  Acros Organics 172580250

Code : 29

Additional Details : CAS Number : 10191-18-1 Weight : 0.08000kg


Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 10082210

  • £36.15 / 25g

In stock, ready to ship: 4

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Biological buffer
Specifications

Chemical Identifiers

10191-18-1
213.25
AJTVSSFTXWNIRG-UHFFFAOYSA-N
73243
2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
C6H15NO5S
MFCD00007533
N, N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid
CHEBI:39045
OCCN(CCO)CCS(O)(=O)=O

Specifications

0.095 max. (1 M in H2O) at 260nm
Solubility in water: soluble.
99.0
(1M in H2O) clear colorless
C6H15NO5S
MFCD00007533
Solubility in water: soluble.
OCCN(CCO)CCS(O)(=O)=O
5ppm max.
73243
213.25
≥99%
Biochemical
Glass bottle
152.0°C to 154.0°C
25g
BES
10191-18-1
100.0
99+%
(HOCH2CH2)2NCH2CH2SO3H
N, N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid
AJTVSSFTXWNIRG-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
213.25
CHEBI:39045
Powder
Authentic
0.5 % max. (100°C, 2 hrs)
0.1 % max.
Beige to Brown
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BES

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates