missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ BES, 0.5M buffer soln., pH 6.5

Product Code. 15413369
Click to view available options
Quantity:
100mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 15413369

missing translation for 'mfr': Alfa Aesar™ J60624.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 66992-27-6
Concentration 0.5M buffer soln.
pH 6.5
Quantity 100mL
Molecular Formula C6H14NNaO5S
MDL Number MFCD00067425
InChI Key CFQLQLSIZOWFNV-UHFFFAOYSA-M
SMILES C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+]
IUPAC Name sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate
Molecular Weight (g/mol) 235.23
PubChem CID 23671640
Chemical Name or Material BES buffer soln.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.