missing translation for 'onlineSavingsMsg'
Learn More

BES, 0.5M buffer soln., pH 6.0, Thermo Scientific™

Product Code. 15432899
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
This item is not returnable. View return policy

Product Code. 15432899

Brand: Thermo Scientific Alfa Aesar J60058.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66992-27-6
Molecular Formula C6H14NNaO5S
Molecular Weight (g/mol) 235.23
MDL Number MFCD00067425
InChI Key CFQLQLSIZOWFNV-UHFFFAOYSA-M
PubChem CID 23671640
IUPAC Name sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate
SMILES C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+]

Specifications

Color Colorless
Concentration 0.5 M
pH 6.0
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material BES buffer soln.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.