missing translation for 'onlineSavingsMsg'
Learn More

Beryllium Standard for ICP/MS - Claritas PPT Grade, SPEX CertiPrep™

Product Code. 15265906 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15265906 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15265906 Supplier SPEX Certiprep Supplier No. CLBE21BY
  • ONLINE EXCLUSIVE PRICE
    £102.00
    £88.29 / 125mL
    Save £13.71
    13% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7440-41-7, 7697-37-2, 7732-18-5
CAS Max % 0.0, 2.0, 100.0
Analyte or Component Names Beryllium Standard
Physical Form Fluid
Molecular Formula Be
MDL Number MFCD00134032
InChI Key ATBAMAFKBVZNFJ-UHFFFAOYSA-N
SMILES [Be]
Element Beryllium Standard
IUPAC Name beryllium
Solvent or Matrix 2% HNO3
Molecular Weight (g/mol) 9.01
PubChem CID 5460467
Quantity 125 mL
ChEBI CHEBI:33783
Grade Claritas PPT
Concentration or Composition (by Analyte or Components) 1mg/L
Chemical Name or Material Beryllium Standard for ICP/MS - Claritas PPT Grade, ICP/MS
Show More Show Less
compliance-icons
Product Identifier
  • 1 µg/mL Beryllium
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.