missing translation for 'onlineSavingsMsg'
Learn More

Beryllium oxide, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11376697
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376697 25 g 25g
11386697 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11376697 Supplier Thermo Scientific Alfa Aesar Supplier No. 010001.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Beryllium oxide is widely used in industry due to its low density and high thermal and electrical conductivity. It is used in high-technology ceramics, electronic heat sinks, electrical insulators, high-power devices, high-density electrical circuits, microwaves, X-ray windows, nuclear reactor fuels and for various aerospace and military applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-56-9
Molecular Formula BeO
Molecular Weight (g/mol) 25.01
MDL Number MFCD00003457
InChI Key LTPBRCUWZOMYOC-UHFFFAOYSA-N
Synonym beryllium oxide, beryllia, bromellete, thermalox, beryllium monoxide, natural bromellite, thermalox 995, ccris 83, beryllium oxide beryllium and beryllium compounds
PubChem CID 14775
ChEBI CHEBI:62842
IUPAC Name oxoberyllium
SMILES [Be]=O

Specifications

Boiling Point 4300°C
Melting Point 2575°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99% (metals basis)
Solubility Information Insoluble in water. Slowly soluble in concentrated acids,alkali hydroxides
Formula Weight 25.01
Density 3.01
Chemical Name or Material Beryllium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.