missing translation for 'onlineSavingsMsg'
Learn More

Beryllium oxide, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. p-7050074
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11353637

Brand: Thermo Scientific Alfa Aesar 035480.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Beryllium oxide is used in rocket engines. It is also used as a structural ceramic for high-performance microwave devices, vacuum tubes, magnetrons, and gas lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-56-9
Molecular Formula BeO
Molecular Weight (g/mol) 25.01
MDL Number MFCD00003457
InChI Key LTPBRCUWZOMYOC-UHFFFAOYSA-N
Synonym beryllium oxide, beryllia, bromellete, thermalox, beryllium monoxide, natural bromellite, thermalox 995, ccris 83, beryllium oxide beryllium and beryllium compounds
PubChem CID 14775
ChEBI CHEBI:62842
IUPAC Name oxoberyllium
SMILES [Be]=O

Specifications

Boiling Point 4300°C
Melting Point 2575°C
Physical Form Powder
Quantity 1 g
UN Number UN1566
Merck Index 14,1172
Formula Weight 25.01
Percent Purity 99.95%
Odor Odorless
Density 3.01g/mL
Assay (metals basis)
Chemical Name or Material Beryllium oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.