missing translation for 'onlineSavingsMsg'
Learn More

Beryllium lump, irregular, 3-38mm (0.1-1.5in), 99.5% (metals basis), Thermo Scientific Chemicals

Catalog Number 11377587
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-41-7
Be
9.01
MFCD00134032
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
5460467
CHEBI:33783
beryllium
[Be]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-41-7
Be
9.01
MFCD00134032
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
5460467
CHEBI:33783
beryllium
[Be]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Appearance Gray irregular lump
Assay Percent Range 99.5% (Metals basis)
Quantity 50 g
Odor Odorless
Form Lump
Chemical Name or Material Beryllium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'