missing translation for 'onlineSavingsMsg'
Learn More

Beryllium AA Standard: Be @ 1000 μg/mL in 5% HNO3, VHG™

Product Code. 30040564
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040564 500 mL 500mL
30040622 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30040564 Supplier VHG Labs Supplier No. VHGAABEN500
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Beryllium AA Standard: Be @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Beryllium
CAS 7440-41-7
Methods AA/GFAA
Analyte or Component Names Beryllium
Shipping Condition Room Temperature
Molecular Formula Be
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Glucinium, Beryllium dust, Beryllium metal, Beryllium powder
InChI Formula InChI=1S/Be
SMILES [Be]
IUPAC Name beryllium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 9.012
Quantity 500 mL
Formula Weight 9.012 g/mol
Show More Show Less
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.