missing translation for 'onlineSavingsMsg'
Learn More

Beryllium, AAS standard solution, Specpure™ Be 1000μg/mL

Product Code. 11396329
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396329 500 mL 500mL
11329149 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11396329 Supplier Thermo Scientific Alfa Aesar Supplier No. 088053.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 19049-40-2 | Be in 5% HNO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19049-40-2 , 7732-18-5
Molecular Formula Be in 5% HNO3
MDL Number MFCD00134032

Specifications

Vapor Pressure 17mm Hg at 20°C
Physical Form Liquid
Color Colorless
Quantity 500 mL
Grade AAS Standard
Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Beryllium oxide acetate: 1.13%; Water: 93.87%
Chemical Name or Material Beryllium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.