missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BERP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583900
This item is not returnable.
View return policy
Description
Immunogen sequence: QHKAALQRQL EAVRGRLPQL SAAIALVGGI SQQLQERKAE ALAQISAAFE DLEQALQQRK QALVSDLETI CGAKQKVLQ Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.
The protein encoded by this gene is a member of the tripartite motif family, also called the 'RING-B-box-coiled-coil' subgroup of RING finger proteins. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to cytoplasmic filaments. It is similar to a rat protein which is a specific partner for the tail domain of myosin V, a class of myosins which are involved in the targeted transport of organelles. The rat protein can also interact with alpha-actinin-4. Thus it is suggested that this human protein may play a role in myosin V-mediated cargo transport. Alternatively spliced transcript variants encoding the same isoform have been identified.
Specifications
| BERP | |
| Polyclonal | |
| Unconjugated | |
| TRIM3 | |
| Berp; BERP1; brain expressed ring finger; brain-expressed RING finger protein; Hac1; RING finger protein 22; RING finger protein 97; RING finger protein HAC1; RNF22; RNF97; Trim3; tripartite motif containing 3; tripartite motif protein 3; tripartite motif protein TRIM3; tripartite motif-containing 3; tripartite motif-containing protein 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10612 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75382 | |
| TRIM3 | |
| Recombinant protein corresponding to Human TRIM3. Recombinant protein control fragment (Product #RP-98384). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction