missing translation for 'onlineSavingsMsg'
Learn More

Bergaptol, MedChemExpress

Product Code. 16431956
Encompass_Preferred
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 16431956

Brand: Medchem Express HY76316

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bergaptol is a hydroxylated psoralen that acts as a potent inhibitors of debenzylation activity of CYP3A4 enzyme with an IC50 value of 24.92 uM.

Bergaptol is a hydroxylated psoralen that acts as a potent inhibitors of debenzylation activity of CYP3A4 enzyme with an IC50 value of 24.92 uM.

Specifications

Chemical Name or Material Bergaptol
CAS 486-60-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C11H6O4
Quantity 500 mg
Synonym 5-Hydroxypsoralen 4-Hydroxybergapten
Solubility Information DMSO : ≥ 50 mg/mL (247.33 mM) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C1C=CC(C(O1)=C2)=C(O)C3=C2OC=C3
Molecular Weight (g/mol) 202.16
Formula Weight 202.16
Percent Purity 98.79%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.