missing translation for 'onlineSavingsMsg'
Learn More

Berberine Chloride, Mikromol™

Product Code. 17908744
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17908744

Brand: Mikromol MM1632.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Berberine Chloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Berberine chloride
CAS 633-65-8
Analyte or Component Names Berberine chloride
Shipping Condition Room Temperature
Molecular Formula C20 H18 N O4 . Cl
Certifications/Compliance ISO 17025
Synonym Benzo[g]-1,3-benzodioxolo[5,6-a]quinolizinium, 5,6-dihydro-9,10-dimethoxy-, chloride, Berberine, chloride, Berbinium, 7,8,13,13a-tetradehydro-9,10-dimethoxy-2,3-(methylenedioxy)-, chloride, Berberine hydrochloride, Berberinium chloride, NSC 163088, NSC 646666
InChI Formula InChI=1S/C20H18NO4.ClH/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2;/h3-4,7-10H,5-6,11H2,1-2H3;1H/q+1;/p-1
SMILES [Cl-].COc1ccc2cc3c4cc5OCOc5cc4CC[n+]3cc2c1OC
IUPAC Name 5,6-Dihydro-9,10-dimethoxybenzo[g]-1,3-benzodioxolo[5,6-a]quinolizinium chloride
Molecular Weight (g/mol) 371.81
Quantity 250 mg
Formula Weight 371.0924 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.