missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Berberine chloride

Product Code. 16407456
Encompass_Preferred
Click to view available options
:
100 mg
This item is not returnable. View return policy

Product Code. 16407456

Brand: MedChemExpress HY18258

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Berberine chloride is an alkaloid that acts as an antibiotic. Berberine chloride induces reactive oxygen species (ROS) generation and inhibits DNA topoisomerase. Antineoplastic properties.

Specifications

Content And Storage 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Molecular Weight (g/mol) 371.81 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 633-65-8
Product Type Natural Products
Purity 98.0%
Solubility DMSO : 12.5 mg/mL (33.62 mM∣Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
Synonym Natural Yellow 18 chloride
Target Topoisomerase∣Autophagy∣Bacterial∣Reactive Oxygen Species∣Antibiotic
Quantity 100 mg
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.