missing translation for 'onlineSavingsMsg'
Learn More

Berbamine dihydrochloride, MedChemExpress

Product Code. 16411516
Encompass_Preferred
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
This item is not returnable. View return policy

Product Code. 16411516

Brand: Medchem Express HYN0714A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Berbamine dihydrochloride is an inhibitor of NF-κB activity with remarkable anti-myeloma efficacy.

Berbamine dihydrochloride is an inhibitor of NF-κB activity with remarkable anti-myeloma efficacy.

Specifications

Chemical Name or Material Berbamine dihydrochloride
CAS 6078-17-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C37H42Cl2N2O6
Quantity 200 mg
Solubility Information DMSO : ≥ 100 mg/mL (146.70 mM)
SMILES COC1=C(OC2=CC([C@]3([H])CC(C=C4)=CC=C4O5)=C(CCN3C)C=C2OC)C([C@@]6([H])CC7=CC=C(O)C5=C7)=C(CCN6C)C=C1OC.Cl.Cl
Molecular Weight (g/mol) 681.65
Formula Weight 681.65
Percent Purity 95.25%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.