missing translation for 'onlineSavingsMsg'
Learn More

Bepridil hydrochloride, MedChemExpress

Product Code. 16433828
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16433828

Brand: Medchem Express HY103315

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bepridil hydrochloride (CERM 1978) is a calcium channel blocker, with antianginal activity.

Bepridil hydrochloride (CERM 1978) is a calcium channel blocker, with antianginal activity.

Specifications

Chemical Name or Material Bepridil hydrochloride
Physical Form Solid
CAS 68099-86-5
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C24H35ClN2O
For Use With (Application) COVID-19-immunoregulation
Synonym CERM 1978
Solubility Information DMSO : ≥ 150 mg/mL (372.21 mM) ∣H2O : 12.5 mg/mL (31.02 mM; Need ultrasonic)
SMILES [H]Cl.CC(C)COCC(N1CCCC1)CN(C2=CC=CC=C2)CC3=CC=CC=C3
Molecular Weight (g/mol) 403.0
Formula Weight 403.0
Percent Purity 99.25%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.