missing translation for 'onlineSavingsMsg'
Learn More

Bepotastine besilate, MedChemExpress

Product Code. 16494147
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494147 50 mg 50mg
16477986 100 mg 100mg
16408268 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16494147 Supplier Medchem Express Supplier No. HYA0015
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Bepotastine besilate is a selective and orally active second-generation histamine H1 receptor antagonist, can suppress the expression of nerve growth factor (NGF). Bepotastine besilate has the potential for allergic rhinitis, allergic conjunctivitis and urticaria/pruritus research.

Bepotastine besilate is a selective and orally active second-generation histamine H1 receptor antagonist, can suppress the expression of nerve growth factor (NGF). Bepotastine besilate has the potential for allergic rhinitis, allergic conjunctivitis and urticaria/pruritus research.

Specifications

Chemical Name or Material Bepotastine besilate
CAS 190786-44-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H31ClN2O6S
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (182.80 mM)
SMILES O=C(CCCN1CCC(CC1)O[C@H](C2=NC=CC=C2)C3=CC=C(C=C3)Cl)O.O=S(C4=CC=CC=C4)(O)=O
Molecular Weight (g/mol) 547.06
Formula Weight 547.06
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.