Learn More
Benzylurea, 98%
Descrizione
Benzylurea is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzylurea is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifica
Specifica
| Melting Point | 146°C to 151°C |
| Quantity | 100 g |
| Beilstein | 2208083 |
| Merck Index | 14,1147 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 150.18 |
| Percent Purity | 98% |
| Chemical Name or Material | Benzylurea |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.