missing translation for 'onlineSavingsMsg'
Learn More

Benzylurea, 98%

Product Code. 11431477
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431477 25 g 25g
11441477 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11431477 Supplier Thermo Scientific Alfa Aesar Supplier No. B24945.14
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 538-32-9 | C8H10N2O | 150.181 g/mol

Benzylurea is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzylurea is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-32-9
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00007951
InChI Key RJNJWHFSKNJCTB-UHFFFAOYSA-N
Synonym 1-benzylurea, n-benzylurea, benzylcarbamide, urea, phenylmethyl, phenylmethylurea, urea, benzyl, cc-pmlsc-dma-p105, amino-n-benzylamide, benzyl urea, benzyl-urea
PubChem CID 10853
IUPAC Name benzylurea
SMILES C1=CC=C(C=C1)CNC(=O)N

Specifications

Melting Point 146°C to 151°C
Quantity 25 g
Beilstein 2208083
Merck Index 14,1147
Solubility Information Slightly soluble in water.
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material Benzylurea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.