missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyltrimethylammonium Tribromide, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Benzyltrimethylammonium Tribromide |
| CAS | 111865-47-5 |
| Molecular Formula | C10H16N.Br3 |
| Quantity | 1 g |
| Synonym | N,N,N-Trimethylbenzenemethanaminium Tribromide |
| InChI Formula | InChI=1S/C10H16N.Br3/c1-11(2,3)9-10-7-5-4-6-8-10;1-3-2/h4-8H,9H2,1-3H3;/q+1;-1 |
| SMILES | C[N+](C)(C)Cc1ccccc1.Br[Br-]Br |
| Molecular Weight (g/mol) | 389.95 |
| Formula Weight | 386.8833 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction