missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium hydroxide, 20% w/w aq. soln., Thermo Scientific™

Product Code. 11308868
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11308868 250 mL 250mL
11328868 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308868 Supplier Thermo Scientific Alfa Aesar Supplier No. 041717.AK

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-85-6
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD00008281
InChI Key NDKBVBUGCNGSJJ-UHFFFAOYSA-M
Synonym benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide
PubChem CID 66854
IUPAC Name benzyl(trimethyl)azanium;hydroxide
SMILES C[N+](C)(C)CC1=CC=CC=C1.[OH-]

Specifications

Color Colorless to Yellow
Odor Amine-like
Linear Formula C6H5CH2N(CH3)3OH
Quantity 250 mL
UN Number UN3267
Sensitivity Air Sensitive
Formula Weight 167.25
Concentration or Composition (by Analyte or Components) 20% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Benzyltrimethylammonium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.