missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium Chloride, TRC

Product Code. 30330334
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330334 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330334 Supplier Toronto Research Chemicals Supplier No. TRCB316173250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyltrimethylammonium Chloride
CAS 56-93-9
Molecular Formula C10H16ClN
Quantity 250 mg
Synonym Benzyltrimethylammonium Chloride, Ammonium, benzyltrimethyl-, chloride (8CI), Benzenemethanaminium, N,N,N-trimethyl-, chloride (9CI), Benzyltrimethylammonium chloride (6CI), Amberlite IRC 178(Cl), BTM, BTMAC 100, BTMAC 50, Hipochem Migrator J, Marcor 716, TMBAC, Trimethylbenzylammonium chloride, Variquat B 200
InChI Formula InChI=1S/C10H16N.ClH/c1-11(2,3)9-10-7-5-4-6-8-10;/h4-8H,9H2,1-3H3;1H/q+1;/p-1
SMILES [Cl-].C[N+](C)(C)Cc1ccccc1
IUPAC Name benzyl(trimethyl)azanium;chloride
Molecular Weight (g/mol) 185.7
Formula Weight 185.0971
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.