missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriethylammonium bromide, 98+%

Product Code. 11472457
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11472457

Brand: Thermo Scientific Alfa Aesar A14291.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5197-95-5 | C13H22BrN | 272.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5197-95-5
Molecular Formula C13H22BrN
Molecular Weight (g/mol) 272.23
MDL Number MFCD00011822
InChI Key CHQVQXZFZHACQQ-UHFFFAOYSA-M
Synonym benzyltriethylammonium bromide, n-benzyl-n,n-diethylethanaminium bromide, benzyl triethyl ammonium bromide, triethylbenzylammonium bromide, bteab, benzenemethanaminium, n,n,n-triethyl-, bromide, benzyltriethyl ammonium bromide, n-benzyl-n,n,n-triethylammoniumbromide, benzyltriethylazanium bromide, pubchem7797
PubChem CID 165294
IUPAC Name benzyl(triethyl)azanium;bromide
SMILES [Br-].CC[N+](CC)(CC)CC1=CC=CC=C1

Specifications

Melting Point ∼200°C (decomposition)
Quantity 500 g
Beilstein 3739113
Sensitivity Hygroscopic
Formula Weight 272.24
Percent Purity ≥98%
Chemical Name or Material Benzyltriethylammonium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.