missing translation for 'onlineSavingsMsg'
Learn More

Benzyltri-n-propylammonium chloride, 96%

Product Code. 15478779
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15478779 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15478779 Supplier Thermo Scientific Alfa Aesar Supplier No. L00675.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5197-87-5 | C16H28ClN | 269.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5197-87-5
Melting Point ∽190°C (decomposition)
Molecular Formula C16H28ClN
Linear Formula C6H5CH2N(CH2CH2CH3)3Cl
MDL Number MFCD00051869
Quantity 10 g
Beilstein 4280671
Sensitivity Hygroscopic
Synonym benzyltripropylammonium chloride, n-benzyl-n,n-dipropylpropan-1-aminium chloride, benzyl tripropyl ammonium chloride, benzyltri-n-propylammonium chloride, benzyltripropylazanium chloride, benzyl tripropyl azanium chloride, acmc-20alov, c16h28n.cl, benzyl-tri-n-propylammonium chloride
InChI Key YTRIOKYQEVFKGU-UHFFFAOYSA-M
SMILES [Cl-].CCC[N+](CCC)(CCC)CC1=CC=CC=C1
IUPAC Name benzyltripropylazanium chloride
Molecular Weight (g/mol) 269.86
PubChem CID 6097067
Formula Weight 269.86
Percent Purity 96%
Chemical Name or Material Benzyltri-n-propylammonium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.