missing translation for 'onlineSavingsMsg'
Learn More

Benzylphosphonic acid, 98%

Product Code. 10511544 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10511544 1 g Glass bottle 1g
10791715 5 g Glass bottle 5g
10258413 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10511544 Supplier Thermo Scientific Acros Supplier No. 393340010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6881-57-8 | C7H7O3P | 170.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6881-57-8
Molecular Formula C7H7O3P
Molecular Weight (g/mol) 170.11
MDL Number MFCD00039519
InChI Key OGBVRMYSNSKIEF-UHFFFAOYSA-L
Synonym benzyl-phosphonic acid, phosphonic acid, phenylmethyl, phenylmethylphosphonic acid, phosphonic acid, p-phenylmethyl, phenyl-methanephosphonic acid anion, acmc-1b2tf, phenylmethanephosphonic acid, alpha-toluenephosphonic acid, benzylphosphonic acid, bmk1-h2
PubChem CID 81312
IUPAC Name benzylphosphonic acid
SMILES [O-]P([O-])(=O)CC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 165.0°C to 166.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CH2P(O)(OH)2
Quantity 1 g
Formula Weight 172.17
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Benzylphosphonic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzylphosphonic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.