missing translation for 'onlineSavingsMsg'
Learn More

Benzyloxyacetic acid, 95%, Thermo Scientific Chemicals

Catalog Number 10727475
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
30379-55-6
C9H10O3
166.18
MFCD00274211
GRZHHTYDZVRPIC-UHFFFAOYSA-N
benzyloxyacetic acid, 2-benzyloxy acetic acid, benzyloxy acetic acid, acetic acid, phenylmethoxy, phenylmethoxyaceticacid, 2-phenylmethoxy acetic acid, benzyloxy-acetic acid, 2-benzyloxyacetic acid, acetic acid, 2-phenylmethoxy
290301
2-phenylmethoxyacetic acid
OC(=O)COCC1=CC=CC=C1
This item is not returnable. View return policy
30379-55-6
C9H10O3
166.18
MFCD00274211
GRZHHTYDZVRPIC-UHFFFAOYSA-N
benzyloxyacetic acid, 2-benzyloxy acetic acid, benzyloxy acetic acid, acetic acid, phenylmethoxy, phenylmethoxyaceticacid, 2-phenylmethoxy acetic acid, benzyloxy-acetic acid, 2-benzyloxyacetic acid, acetic acid, 2-phenylmethoxy
290301
2-phenylmethoxyacetic acid
OC(=O)COCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.1620g/mL
Boiling Point 137°C to 139°C (0.6 mmHg)
Flash Point 113°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2OCH2COOH
Quantity 1 g
Specific Gravity 1.162
Formula Weight 166.18
Percent Purity 95%
Chemical Name or Material Benzyloxyacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • Benzyloxyacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'